Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MITF Antibody (SPM290), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311527H
Description
MITF Monoclonal antibody specifically detects MITF in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MITF | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
4286 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
SPM290 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
BHLHE32, bHLHe32MI, Class E basic helix-loop-helix protein 32, microphthalmia-associated transcription factor, Waardenburg syndrome, type 2A, WS2A | |
NH2 terminus fragment of human MITF (Uniprot: O75030) | |
0.1 mL | |
Primary | |
Human, Mouse (Negative), Rat (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction