Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MITF Mouse anti-Human, Clone: PCRP-MITF-1D9, Novus Biologicals™

Mouse Monoclonal Antibody
$251.50 - $504.00
Specifications
Antigen | MITF |
---|---|
Clone | PCRP-MITF-1D9 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Peptide Array |
Description
MITF Monoclonal antibody specifically detects MITF in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
MITF | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
O75030 | |
4286 | |
IgG2b | |
Protein A or G purified |
PCRP-MITF-1D9 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
BHLHE32, bHLHe32MI, Class E basic helix-loop-helix protein 32, microphthalmia-associated transcription factor, Waardenburg syndrome, type 2A, WS2A | |
Recombinant full-length human MITF protein (Uniprot: O75030) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title