Learn More
MITF Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™

Description
Specifications
Specifications
| Antigen | MITF |
| Applications | ChIP sequencing (ChIP-seq) |
| Classification | Monoclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 140mM HEPES, 35mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 |
| Gene | MITF |
| Gene Accession No. | O75030 |
| Gene Alias | BCC2; BHLHE32; black eyed white; BOS_20839; Bw; Class E basic helix-loop-helix protein 32; CMM8; Gsfbcc2; melanocyte inducing transcription factor; melanogenesis associated transcription factor; Mi; microphtalmia-associated transcription factor; microphthalmia transcription factor; Microphthalmia-associated transcription factor; microphthalmia-associated transcription factor A; microphthalmia-associated transcription factor isoform H protein; microphthalmia-associated transcription factor Trnc-1; microphthalmia-associated transcription factor Trnc-2; microphthalmia-associated transcription factor variant 1; microphthalmia-associated transcription factor variant 2; microphthalmia-associated transcription factor variant 3; microphthalmia-associated transcription factor variant MITF-M(+); MITF; MITF-H; MITF-M; transcription factor, isoform 7; Vit; Vitiligo; Wh; WS2; WS2A |
| Gene Symbols | MITF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.