Learn More
Invitrogen™ MITFA Polyclonal Antibody

Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. Positive control: zebrafish muscle.
Specifications
Specifications
| Antigen | MITFA |
| Applications | Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene Alias | melanocyte inducing transcription factor a; nacre; z3A.1 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of zebrafish Mitfa. The exact sequence is proprietary. |
| Purification Method | Antigen Affinity Chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.