Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MitoBoronic acid
SDP

Catalog No. 794950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-495-0 50 mg
79-491-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 79-495-0 Supplier Tocris Bioscience™ Supplier No. 7949/50
Add to Cart
Add to Cart

Ratiometric mass spectrometry mitochondrial hydrogen peroxide (H2O2) probe

MitoBoronic acid (MitoB) is a ratiometric mass spectrometry mitochondrial hydrogen peroxide (H2O2) probe. MitoBoronic acid is taken up into cells and accumulates inside the mitochondrial matrix where it reacts to H2O2 to be converted in MitoP. Measurement of MitoB/MitoP ratio by liquid chromatography tandem mass spectrometry (LC-MS/MS) is an indicator of the mitochondrial matrix H2O2 concentration. This method can be used in vitro and in vivo.

Specifications

Chemical Name or Material [(3-Boronophenyl)methyl]triphenylphosphonium bromide
CAS 1247025-84-8
Quantity 50 mg
Target Additional ROS Donor Products
Molecular Formula C25H23BBrO2P
Product Type Bioactive Small Molecule
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.