Learn More
Description
MitoBrilliant™ 646 is a next-generation universal stain for mitochondria in both live and fixed cells. It is retained in mitochondria following fixation with exceptionally clear staining and is retained in the mitochondria of live cells following loss of the mitochondrial membrane potential. Additionally, MitoBrilliant™ 646 can be used to stain pre-fixed cells/tissue, making this dye suitable for IHC/ICC.
Two dyes that accumulate in the mitochondria of live cells in a mitochondrial membrane potential (Δψm) dependent manner are also available: MitoBrilliant™ Live 646 (red emission) and MitoBrilliant™ Live 549 (yellow/orange emission).
Please refer to the protocol for guidelines on product use, and download the MitoBrilliant Product Guide to view data for each of the dyes in different applications.
MitoBrilliant is a trademark of Bio-Techne Corp.
Specifications
Specifications
| Content And Storage | Store at -20°C |
| Quantity | 50 μg (Trial size) |
| Product Line | Tocris |
| Target | Mitochondria Dyes and Probes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
