Learn More
Description
The MitoBrilliant™ Live dyes accumulate in the mitochondria of live cells in a mitochondrial membrane potential (Δψm) dependent manner. Upon loss of the mitochondrial membrane potential, the dyes disperse, and so provide a dynamic assessment of the mitochondrial membrane potential in live cells. Two dyes are available: MitoBrilliant™ Live 646 (red emission) and MitoBrilliant™ Live 549 (yellow/orange emission).
A universal stain for mitochondria in both live and fixed cells is also available: MitoBrilliant™ 646 .
Please refer to the protocol for guidelines on product use, and download the MitoBrilliant Product Guide to view data for each of the dyes in different applications.
MitoBrilliant is a trademark of Bio-Techne Corp.
Specifications
Specifications
| Quantity | 50 μg (Trial size) |
| Product Line | Tocris |
| Target | Mitochondria Dyes and Probes |
| Purity | 95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
