Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mitochondria Antibody (MTC719), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234687PEATT594
Description
Mitochondria Monoclonal specifically detects Mitochondria in Human samples. It is validated for Flow Cytometry.Specifications
Mitochondria | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
Mitochondrial fraction of HeLa cells | |
0.1 mL | |
This monoclonal antibody recognizes a 60kDa antigen associated with the mitochondria in cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the mitochondria in cells from a wide variety of animals, but not insects and bacteria. It can be used to stain the mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-like pattern in normal and malignant cells and may be used to stain mitochondria of cells in fixed or frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
MTC719 | |
Flow Cytometry | |
Mouse | |
Protein A or G purified | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction