Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mitochondria Membrane Integrity Antibody Cocktail
GREENER_CHOICE

Catalog No. 457799
Encompass_Preferred
Change view
Click to view available options
Quantity:
255 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-779-9 255 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-779-9 Supplier Invitrogen™ Supplier No. 457799
Add to Cart
Add to Cart

Antibody Cocktail

Mitochondria Membrane Integrity cocktail of monoclonal antibodies specific to human, bovine, mouse, rat mitochondria membranes. This antibody is validated for use in Western Blot, and can be used to assess mitochondrial membrane integrity during and after purification. Contains: 50 μg of Porin clone #2B12AF12 - Human VDAC1. Concentration 2 μg/mL. 50 μg of Cyclophilin D clone #E11AE12BD4 - Rat CypD/PPIF. Concentration 2 μg/mL. 50 μg of C-III-Core 1 clone # 16D10AD9AH5 - Bovine Heart Complex II/III. Concentration 2 μg/mL. 5 μg of Cytochrome C clone #37BA11 - Human Cytochrome c. Concentration 0.1 μg/mL. 100 μg of C-V-a clone #7H10BD4F9 - Bovine Complex V. Concentration 4 μg/mL. This antibody cocktail (2.37 mg/mL) should be diluted to a final working concentration of 10.2 μg/mL for use in Western blot applications.

Voltage-dependent anion channel (VDAC) proteins are abundant, pore-forming proteins belonging to the eukaryotic mitochondrial porins. It was discovered in the mitochondrial outer membrane. It also is expressed in the plasma membrane. At least three different VDAC genes have been identified in vertebrates. VDAC proteins are known to play an essential role in cellular metabolism and in the early stages of apoptosis. For example, VDAC contitutes a major pathway by which metabolites such as ADP/ATP, succinate and citrate are exchanged between the cytosol and mitochondria. And VDAC1 in the plasma membrane establishes a novel level of apoptosis regulation putatively via its redox activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mitochondria Membrane Integrity
Applications Western Blot
Classification Cocktail
Clone Cocktail
Concentration 2.37 mg/mL
Conjugate Unconjugated
Formulation HEPES buffered saline with 0.02% sodium azide
Gene CYC1
Gene Accession No. P00125, P08574, P21796, P29117, P30404, P30405, P45879, P62894, P62897, P62898, P99999, Q60932, Q99KR7, Q9D0M3, Q9Z2L0
Gene Alias 16D10AD9AH5; 20B12AF2; 2610002H19Rik; 37BA11; 7H10BD4F9; AA408921; AL033343; AW457192; BR1-VDAC; brain-derived voltage-dependent anion channel 1; Complex III subunit 4; complex III subunit IV; CYC; Cyc1; cyclophilin 3; cyclophilin D; Cyclophilin F; Cycs; CYCSA; CYP3; CypD; CyP-D; CyP-F; CyP-M; cytc; Cytochrome b-c1 complex subunit 4; cytochrome c; Cytochrome C, expressed in somatic tissues; cytochrome c, somatic; cytochrome c1; Cytochrome c-1; cytochrome c1, heme protein, mitochondrial; E11AE12BD4; HCS; MC3DN6; MGC128356 protein; mitochondrial cyclophilin; mitochondrial Cyclophilin D; mVDAC1; mVDAC5; outer mitochondrial membrane protein porin 1; Peptidyl-prolyl cis-trans isomerase F, mitochondrial; peptidyl-prolyl cis-trans isomerase, mitochondrial; peptidylprolyl isomerase F; peptidylprolyl isomerase F (cyclophilin F); plasmalemmal porin; PORIN; Porin 31HL; porin 31HM; PPIase; PPIase F; PPIF; Rotamase F; rVDAC1; THC4; ubiquinol-cytochrome-c reductase complex cytochrome c1 subunit; Unknown (protein for MGC:134266); UQCR4; VDAC; Vdac1; VDAC-1; Vdac5; VDAC-5; voltage dependent anion channel 1; voltage-dependent anion channel 1; voltage-dependent anion-selective channel protein 1; Voltage-dependent anion-selective channel protein 5
Gene Symbols CYC1, Cycs, PPIF, Vdac1
Host Species Mouse
Purification Method IgG fraction
Quantity 255 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10105, 105675, 13063, 1537, 22333, 25309, 282119, 282819, 300047, 414346, 510767, 512500, 54205, 66445, 7416, 83529
Target Species Human, Mouse, Rat, Bovine
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype Ig
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.