Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mitochondria Monoclonal Antibody (113-1)
GREENER_CHOICE

Catalog No. PIMA547691
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547691 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547691 Supplier Invitrogen™ Supplier No. MA547691
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specific for endogenous levels of the ~60 kDa human mitochondria. The antibody works on acetone fixed and 2% paraformaldehyde fixed cells. The antibody also works for immunohistochemistry on 2% paraformaldehyde fixed (5-10 minutes) cryostat sections.

Antibodies that recognize subcellular organelles or compartments of human cells are useful in identification of these organelles in cells, tissues, and biochemical preparations. Anti-mitochondria antibodies can be used to stain mitochondria in cell/tissue preparations and can be used as a marker in subcellular fractions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mitochondria
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 113-1
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene EWSR1
Gene Accession No. Q01844, Q61545
Host Species Mouse
Immunogen Human cell homogenate.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14030, 2130, 609786
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.