Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitochondria Rabbit anti-Human, Clone: MTC02/2860R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$304.50

Specifications

Antigen Mitochondria
Clone MTC02/2860R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122092
SDP
missing translation for 'viewDocuments'
Novus Biologicals
NBP30730920UG
20 μg
Each of 1 for $304.50
Only null left
Add to Cart
 
Description

Description

Mitochondria Monoclonal specifically detects Mitochondria in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Mitochondria
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
Semi-Purified mitochondrial preparation
Primary
Store at 4°C.
MTC02/2860R
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
IgG
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.