Learn More
MilliporeSigma™ Mitochondria, surface of intact mitochondria, Clone: 113-1,™ (Chemicon™)
Supplier:  MilliporeSigma™ MAB1273/DEL
Encompass
Catalog No. 50-174-784
Description
Description
This antibody recognizes a 60kDa nonglycosylated protein component of mitochondria found in human cells. It can be used to stain mitochondria in cell, tissure or biochemical preparations and can be used as a marker of the mitochondria in subcellular fractions.
Kehat, I. et al.(2004). Electromechanical integration of cardiomyocytes derived from human embryonic stem cells.Nat Biotechnol. 22: 1282-1289
Immunohistochemistry, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mitochondria, surface of intact mitochondria
Unconjugated
100 μL
Human
IgG1
Monoclonal
Mouse
Primary
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.