Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitochondrial ribosomal protein L11 Antibody, DyLight 350, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP306019UV

Catalog No. NB145911


Only null left
Add to Cart

Description

Description

Mitochondrial ribosomal protein L11 Polyclonal specifically detects Mitochondrial ribosomal protein L11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Mitochondrial ribosomal protein L11
Polyclonal
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
L11MT, MGC111024, mitochondrial ribosomal protein L11, MRP-L11,39S ribosomal protein L11, mitochondrial
Produced in rabbits immunized with E. coli-derived Human Mitochondrial ribosomal protein L11 fragment.
0.1 mL
Core ESC Like Genes, Stem Cell Markers
65003
Store at 4°C in the dark.
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 350
50mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.