Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mitochondrial ribosomal protein L11 Antibody (OTI1E5), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272707AF750
Description
Mitochondrial ribosomal protein L11 Monoclonal specifically detects Mitochondrial ribosomal protein L11 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Mitochondrial ribosomal protein L11 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
MRPL11 | |
Full length recombinant protein of human Mitochondrial ribosomal protein L11 (NP_057134) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1E5 | |
Western Blot, Immunohistochemistry | |
L11MT, MGC111024, mitochondrial ribosomal protein L11, MRP-L11,39S ribosomal protein L11, mitochondrial | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
65003.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction