Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mitochondrial ribosomal protein L30 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30329520UL
Description
mitochondrial ribosomal protein L30 Polyclonal specifically detects mitochondrial ribosomal protein L30 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| mitochondrial ribosomal protein L30 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51263 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 39S ribosomal protein L30, mitochondrial, L28mt, L30mt, mitochondrial ribosomal protein L30, MRP-L28FLJ44438, MRPL28MGC3314,39S ribosomal protein L28, mitochondrial, MRP-L30, RPML28MGC24095 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-161 of human mitochondrial ribosomal protein L30 (NP_660213.1). | |
| 20 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction