Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mitocondrial Translational Initiation Factor 3 Antibody (OTI4B6), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272708AF532
Description
Mitocondrial Translational Initiation Factor 3 Monoclonal specifically detects Mitocondrial Translational Initiation Factor 3 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Mitocondrial Translational Initiation Factor 3 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
MTIF3 | |
Recombinant protein fragment corresponding to amino acids 56-278 of human Mitocondrial Translational Initiation Factor 3 (NP_690876) produced in E.coli. | |
0.1 mL | |
219402.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4B6 | |
Western Blot, Immunohistochemistry | |
FLJ33676, IF3(mt), IF-3mt, mitochondrial, mitochondrial translational initiation factor 3 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Canine, Monkey | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction