Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mitocondrial Translational Initiation Factor 3 Antibody (OTI4B6) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272708
Description
Mitocondrial Translational Initiation Factor 3 Monoclonal specifically detects Mitocondrial Translational Initiation Factor 3 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Mitocondrial Translational Initiation Factor 3 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
FLJ33676, IF3(mt), IF-3mt, mitochondrial, mitochondrial translational initiation factor 3 | |
Mouse | |
31.5 kDa | |
100 μg | |
219402 | |
Human, Canine, Monkey | |
Lyophilized |
Western Blot, Immunohistochemistry | |
OTI4B6 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
MTIF3 | |
Human recombinant protein fragment corresponding to amino acids 56-278 of human MTIF3 (NP_690876) produced in E.coli. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction