Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mitocondrial Translational Initiation Factor 3 Antibody (OTI4B6), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246034
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
Mitocondrial Translational Initiation Factor 3 Monoclonal specifically detects Mitocondrial Translational Initiation Factor 3 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Mitocondrial Translational Initiation Factor 3 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
FLJ33676, IF3(mt), IF-3mt, mitochondrial, mitochondrial translational initiation factor 3 | |
Mouse | |
31.5 kDa | |
0.1 mL | |
Primary | |
Human, Canine, Monkey | |
Purified |
Western Blot, Immunohistochemistry | |
OTI4B6 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
MTIF3 | |
Human recombinant protein fragment corresponding to amino acids 56-278 of human MTIF3 (NP_690876) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
219402 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction