Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mitoferrin 2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS7157R

Description
Mitochondrial iron transporter that mediates iron uptake. Probably required for heme synthesis of hemoproteins and Fe-S cluster assembly in non-erythroid cells. The iron delivered into the mitochondria, presumably as Fe(2+), is then probably delivered to ferrochelatase to catalyze Fe(2+) incorporation into protoprophyrin IX to make heme.Specifications
Mitoferrin 2 | |
Polyclonal | |
Unconjugated | |
SLC25A28 | |
2210403D18Rik; BOS_23457; DKFZp547C109; hMRS3/4; LOC100342228; MFRN2; Mitochondrial iron transporter 2; mitochondrial RNA splicing protein 3/4; Mitochondrial RNA-splicing protein 3/4 homolog; mitoferrin 2; Mitoferrin-2; MRS3/4; MRS4L; NPD016; putative mitochondrial solute carrier; SLC25A28; solute carrier family 25 (mitochondrial iron transporter), member 28; Solute carrier family 25 member 28; solute carrier family 25, member 28 | |
Rabbit | |
Protein A | |
RUO | |
81894 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96A46 | |
SLC25A28 | |
KLH conjugated synthetic peptide derived from human Mitoferrin 2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction