Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitofilin Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 16 publications

$222.20 - $537.28

Specifications

Antigen Mitofilin
Dilution Western Blot 1:2000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200-1:500
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1001919SS
SDP
View Documents
Novus Biologicals
NB1001919SS
0.025 mL
Each for $222.20
Only null left
Add to Cart
 
NB1001919
SDP
View Documents
Novus Biologicals
NB1001919
0.1 mL
Each for $537.28
Only null left
Add to Cart
 
Description

Description

Mitofilin Polyclonal specifically detects Mitofilin in Human, Mouse, Rat, Canine, Feline, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Mitofilin
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cellular Markers, Lipid and Metabolism, Mitochondrial Markers, Signal Transduction
Whole antisera with 0.05% Sodium Azide
DKFZp779P1653, inner membrane protein, mitochondrial, MGC111146, Mic 60, Mic60, MICOS Complex Subunit MIC60, mitochondrial (mitofilin), mitofilin, motor protein, p87/89, proliferation-inducing gene 4
IMMT
IgG
Unpurified
Human, mouse, rat, canine and feline. No other species have been tested.
Western Blot 1:2000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200-1:500
Polyclonal
Antisera
RUO
Human, Mouse, Rat, Canine, Feline
Q16891
10989
Whole human recombinant Mitofilin [UniProt# Q16891]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.