Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mitofilin Monoclonal Antibody (MM#7A-2E4AD5), Alexa Fluor™ 488
GREENER_CHOICE

Catalog No. PIMA4564A48
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA4564A48 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA4564A48 Supplier Invitrogen™ Supplier No. MA456400A488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mitofilin Monoclonal Antibody for ICC/IF

Mitofilin is a transmembrane protein of the inner mitochondrial membrane and it is associated with a large multimeric protein complex of about 1200 kDa. Mitofilin has critical functions in mitochondrial morphology and mitochondrial fusion and fission, specifically in the formation of tubular cristae and cristae junctions. Abnormal mitochondrial morphology has been implicated in many human diseases such as myopathy, cardiomyopathy, rhabdomyosarcoma and Whartin's tumor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mitofilin
Applications Immunocytochemistry
Classification Monoclonal
Clone MM#7A-2E4AD5
Concentration 1 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene IMMT
Gene Accession No. Q16891
Gene Alias 1700082C19Rik; cell proliferation-inducing gene 4/52 protein; cell proliferation-inducing protein 52; D830041H16Rik; EMBL:AAI05842.1, ECO:0000312; EMBL:AAI05842.1}; HMP; Immt; immt {ECO:0000312; IMMT antibody; inner membrane mitochondrial protein; inner membrane protein, mitochondrial; inner membrane protein, mitochondrial (mitofilin); MGC133830 protein; MIC60; MICOS complex subunit Mic60; MINOS2; mitochondrial inner membrane organizing system 2; mitochondrial inner membrane protein; mitochondrial inner membrane protein {ECO:0000250; Mitofilin; mitofilin {ECO:0000250; motor protein; P87; p87/89; p87/89 antibody; P89; PIG4; PIG52; proliferation-inducing gene 4; RGD:1310684}; UniProtKB:Q8CAQ8, ECO:0000312; UniProtKB:Q8CAQ8}
Gene Symbols IMMT
Host Species Mouse
Immunogen Human heart mitochondria.
Purification Method Purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10989
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.