Learn More
Invitrogen™ Mitofilin Polyclonal Antibody

Description
Positive Samples: SW480, SW620, H460, HepG2, Mouse spleen, Mouse testis; Cellular Location: Mitochondrion, Mitochondrion inner membrane, Single-pass membrane protein Immunogen sequence: EELEKMKSVI ENAKKKEVAG AKPHITAAEG KLHNMIVDLD NVVKKVQAAQ SEAKVVSQYH ELVVQARDDF KRELDSITPE VLPGWKGMSV SDLADKLSTD D.
Specifications
Specifications
| Antigen | Mitofilin |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.64 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | IMMT |
| Gene Accession No. | Q16891, Q3KR86, Q8CAQ8 |
| Gene Alias | 1700082C19Rik; cell proliferation-inducing gene 4/52 protein; cell proliferation-inducing protein 52; D830041H16Rik; EMBL:AAI05842.1, ECO:0000312; EMBL:AAI05842.1}; HMP; Immt; immt {ECO:0000312; IMMT antibody; inner membrane mitochondrial protein; inner membrane protein, mitochondrial; inner membrane protein, mitochondrial (mitofilin); MGC133830 protein; MIC60; MICOS complex subunit Mic60; MINOS2; mitochondrial inner membrane organizing system 2; mitochondrial inner membrane protein; mitochondrial inner membrane protein {ECO:0000250; Mitofilin; mitofilin {ECO:0000250; motor protein; P87; p87/89; p87/89 antibody; P89; PIG4; PIG52; proliferation-inducing gene 4; RGD:1310684}; UniProtKB:Q8CAQ8, ECO:0000312; UniProtKB:Q8CAQ8} |
| Gene Symbols | IMMT |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.