Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mitofusin-2 Monoclonal Antibody (S153-5), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 56557100UG
This item is not returnable.
View return policy
Description
Mitofusin-2 Monoclonal antibody specifically detects Mitofusin-2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| Mitofusin-2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.4 | |
| O95140, Q80U63, Q8R500 | |
| Mouse | |
| Protein A | |
| RUO | |
| 170731, 64476, 9927 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| S153-5 | |
| Unconjugated | |
| MFN2 | |
| CMT2A; CMT2A2; CPRP 1; CPRP1; D630023P19Rik; Fzo; HMSN6A; HSG; HSG protein; hyperplasia suppressor; hypertension related protein 1; hypertension-related protein; hypertension-related protein 1; Kiaa0214; LOW QUALITY PROTEIN: mitofusin-2; Marf; MFN 2; MFN2; mg:cb01g09; Mitochondrial assembly regulatory factor; mitochondrial transmembrane GTPase FZO1A; mitofusin 2; Mitofusin2; mitofusin-2; si:dkeyp-104h9.2; Transmembrane GTPase MFN2; wu:fb79a11 | |
| Fusion protein corresponding to aa 370-600 (cytoplasmic N-terminus) of mouse Mitofusin-2. This sequence is 97 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction