Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitomycin C, PROMO, Thermo Scientific Chemicals

Catalog No. J63193MAPM
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Mitomycin C is a methylazirinopyrroloindoledione compound showing antineoplastic and antibiotic activities. It is active against a broad-spectrum of gram-positive, gram-negative, and acid-fast bacilli bacteria.

General Description

  • Mitomycin C is a methylazirinopyrroloindoledione compound isolated from the bacterium Streptomyces caespitosus, and other Streptomyces bacterial species that has antineoplastic and antibiotic activities
  • This compound acts as an inhibitor of DNA synthesis, nuclear division, as well as an apoptosis inducer. It can create oxygen radicals, and produce inter-strand DNA crosslinks, inhibiting DNA synthesis
  • Mitomycin C shows alkylating activity and mainly targets the guanine nucleoside sequence 5'-CpG-3'. It inhibits DNA synthesis by covalently reacting with DNA, forming crosslinks between complementary strands of DNA. Consequently, this interaction prevents the separation of complementary DNA strands, inhibiting DNA replication

Application

  • Mitomycin C is active against a broad-spectrum of gram-positive, gram-negative, and acid-fast bacilli bacteria
  • Mitomycin C has strong antineoplastic activity, mainly against Ehrlich ascites tumor cell lines
  • This compound can inhibit B and T cells and macrophage proliferation. It can also impair antigen presentation, as well as the secretion of interferon-γ, TNF- α, and IL-2
  • This compound is used to generate mitotically inactive feeder cells in cell culture systems, such as the mitotically inactive fibroblasts used in embryonic stem cell systems
  • It has been used to treat basal-like cancer cell line in vitro experiments
  • It is particularly toxic to hypoxic cells inhibiting RNA and protein synthesis at high concentrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-07-7
Molecular Formula C15H18N4O5
Molecular Weight (g/mol) 334.33 g/mol
MDL Number MFCD00078109
InChI Key NWIBSHFKIJFRCO-WUDYKRTCSA-N
PubChem CID 5746
ChEBI CHEBI:27504
SMILES CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N

Specifications

Type Mitomycin C
Physical Form Powder
Melting Point 360°C
Color Blue to Purple
Quantity 10 mg
UN Number UN2811
Beilstein 7231816
Merck Index 14,6215
Solubility Information Soluble in methanol, water and dimethyl sulfoxide.
Chemical Name or Material Mitomycin C
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-008-6
RTECSNumber CN0700000
TSCA Yes
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.