Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neomycin Sulfate USP, Medisca™
SDP

Catalog No. 18048076 Shop All Medisca Products
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
1 kg

Neomycin Sulfate USP is a white to slightly yellow powder, or cryodesiccated solid. It is odorless or practically so, and is hygroscopic.

Chemical Identifiers

CAS 1405-10-3

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder/Solid
Color White to Light Yellow
Quantity 500 g
Percent Purity ≥ 600 μg of Neomycin/mg
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.