Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MitoProbe™ TMRM Assay Kit for Flow Cytometry

Catalog No. M20036
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20036 100 assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20036 Supplier Thermo Scientific™ Supplier No. M20036
  • $310.00
    $297.65 / Each of 1

    Save $12.35

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MitoProbe TMRM Assay Kit contains tetramethylrhodamine methyl ester (TMRM) for the detection of mitochondrial membrane potential state and CCCP for the induction of mitochondrial membrane depolarization, if desired.

• Study mitochondrial membrane potential in live cells
• Quick, easy protocol

Determining the state of mitochondrial health can be achieved by detecting changes in the mitochondrial membrane potential. In a healthy cell with active mitochondria, TMRM is readily sequestered, thereby emitting a red-orange fluorescent signal. When apoptosis is induced or a treatment like CCCP is applied, the mitochondrial membrane is depolarized and the TMRM signal diminishes.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage
  • 1 vial TMRM (100 tests)
  • 1 vial 50 mM CCCP
  • Store at -5°C to -30°C.
Detection Method Fluorescence
For Use With (Application) TMRM Assay
For Use With (Equipment) Flow Cytometer
Product Type Flow Cytometry Assay Kit
Format Vial(s)
Quantity 100 assays
Shipping Condition Wet Ice
Emission Wavelength 561
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.