Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Mitosis Manipulatives Kit
SDPSCI_ED

Catalog No. S90331 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
30 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S90331 30 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90331 Supplier United Scientific Supplies Supplier No. MITKIT2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Printed manual and downloadable resources

Instructive and reusable.

Manipulatives make it easier to teach an abstract concept like mitosis in a science classroom. 

  • Teacher demonstration model includes large pieces with magnets attached
  • Five sets of smaller manipulatives for use by students working in groups
  • Reusable

 

Specifications

Grade 6 to 12
For Use With (Application) Helps students in understanding of the process of mitosis.
Includes Oversized teacher manipulatives, magnets for teacher's manipulatives, 5 sets of smaller student manipulatives, printed manual and downloadable resources
Class Size 30 Students
Product Type Mitosis Manipulatives Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.