Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mitotic Cells Monoclonal Antibody (8B3G)
GREENER_CHOICE

Catalog No. MA106200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA106200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA106200 Supplier Invitrogen™ Supplier No. MA106200
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-06200 detects Mitotic cells in human samples. MA1-06200 has sucessfully been used in immunocytochemistry and FACS procedures. The MA1-06200 immunogen is total cell lysate of the human bladder carcinoma cell line T24.

The life cycle of a eukaryotic cell consists of various phases, two of which can morphologically and biochemically be identified. Firstly, during mitosis (M-phase), in which the cell divides into two identical daughter cells, chromosome condensation and spindle formation are microscopically visible. Secondly, in S-phase the DNA of a cell is replicated, a process that can be detected using biochemical techniques, such as the BrdU incorporation assay. In between the M- and S-phase two gap phases occur: the G1-phase, the gap between mitosis and the start of DNA replication, and G2-phase, the gap between completion of DNA replication and the onset of mitosis. From G1-phase a cell can leave the cell cycle and enter G0, a quiescent phase. Regulation of the cell cycle predominantly occurs at three major control points, which govern the transition from G0 to G1, from G1 to S, and from G2 to M-phase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mitotic Cells
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 8B3G
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Host Species Mouse
Immunogen Total cell lysate of the human bladder carcinoma cell line T24.
Purification Method Ammonium sulfate precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.