Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mixed Bed Exchange Resin, IONAC™ NM-60 H+/OH- Form, Type I, Strong acid/strong base; sulfonated/alkyl quaternary ammonium polystyrenes, Beads, 16 to 50 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003298 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-003-298 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-298 Supplier Avantor Supplier No. 463101
Add to Cart
Add to Cart

High quality chemicals for laboratory and specialized industrial use

Specifications

Chemical Name or Material Mixed Bed Exchange Resin, IONAC™ NM-60 H+/OH- Form, Type I
Specific Gravity 1 L = 1.20 kg
Quantity 500 g
Name Note Strong acid/strong base; sulfonated/alkyl quaternary ammonium polystyrenes
Mesh Size 16 to 50 Mesh
Packaging Wide Mouth Amber Glass Bottle
Physical Form Beads
Grade BAKER ANALYZED™ Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.