Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mixed Calibration Standard 1A, ICP, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
100 mL
500 mL
Specifications
Specifications
Chemical Name or Material | Mixed Calibration Standard 1A |
Packaging | Amber Poly Bottle |
CAS Min % | 91.0, 6.65, 0.02, 0.02, 0.01, 0.01 |
CAS Max % | 95.0, 6.92, 0.02, 0.02, 0.01, 0.01 |
CAS | 7647-01-0, 471-34-1, 1327-53-3, 10043-35-3, 7697-37-2, 7732-18-5 |
Odor | Odorless |
Color | Colorless |
Melting Point | Approximately 0°C |
Quantity | 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction