Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mixed Calibration Standard 1A, ICP, Ricca Chemical
SDP

Catalog No. RMXCL1A500 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
100 mL
500 mL

Specifications

Chemical Name or Material Mixed Calibration Standard 1A
Packaging Amber Poly Bottle
CAS Min % 91.0, 6.65, 0.02, 0.02, 0.01, 0.01
CAS Max % 95.0, 6.92, 0.02, 0.02, 0.01, 0.01
CAS 7647-01-0, 471-34-1, 1327-53-3, 10043-35-3, 7697-37-2, 7732-18-5
Odor Odorless
Color Colorless
Melting Point Approximately 0°C
Quantity 500 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.