Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIXL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567818
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567818 Supplier Invitrogen™ Supplier No. PA567818
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Human samples.

MIXL1 is a novel human Mix-like homeobox gene. In normal hematopoiesis, its expression appears to be restricted to immature B- and T-lymphoid cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIXL1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MIXL1
Gene Accession No. Q9H2W2, Q9WUI0
Gene Alias hMix; Homeobox protein MIXL1; homeodomain protein MIX; MILD1; MIX; Mix paired-like homeobox; Mix paired-like homeobox 1; mix.1 homeobox-like protein; Mix1 homeobox-like 1 (Xenopus laevis); MIX1 homeobox-like protein 1; MIXL; Mixl1; Mix-like homeobox protein 1; Mm1; mMix; Mml
Gene Symbols MIXL1
Host Species Rabbit
Immunogen Synthesized peptide derived from human MIXL1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27217, 83881
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.