Learn More
Miz-1/ZBTB17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Miz-1/ZBTB17 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | Miz-1, MIZ1, Myc-interacting zinc finger protein 1, pHZ-67, zinc finger and BTB domain containing 17, zinc finger and BTB domain-containing protein 17, Zinc finger protein 151, zinc finger protein 151 (pHZ-67), Zinc finger protein 60MIZ-1, ZNF151Myc-interacting Zn finger protein-1, ZNF60 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 121-360 of human Miz-1/ZBTB17 (NP_003434.2). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.