Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Miz-1/ZBTB17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB092984 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092984 20 μL
NB092983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092984 Supplier Novus Biologicals Supplier No. NBP30335820UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Miz-1/ZBTB17 Polyclonal specifically detects Miz-1/ZBTB17 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Miz-1/ZBTB17
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Miz-1, MIZ1, Myc-interacting zinc finger protein 1, pHZ-67, zinc finger and BTB domain containing 17, zinc finger and BTB domain-containing protein 17, Zinc finger protein 151, zinc finger protein 151 (pHZ-67), Zinc finger protein 60MIZ-1, ZNF151Myc-interacting Zn finger protein-1, ZNF60
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 121-360 of human Miz-1/ZBTB17 (NP_003434.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7709
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.