Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mizoribine 98.0+%, TCI America™
SDP

Catalog No. M2399250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2399250MG 250 mg
M239950MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2399250MG Supplier TCI America Supplier No. M2399250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50924-49-7
Molecular Formula C9H13N3O6
Molecular Weight (g/mol) 259.22
MDL Number MFCD00057221
InChI Key HZQDCMWJEBCWBR-UHFFFAOYNA-N
Synonym 1-(beta-D-Ribofuranosyl)-5-hydroxyimidazole-4-carboxamide
PubChem CID 104762
IUPAC Name 1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-hydroxy-1H-imidazole-4-carboxamide
SMILES NC(=O)C1=C(O)N(C=N1)C1OC(CO)C(O)C1O

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 250 mg
Formula Weight 259.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Mizoribine
RTECSNumber NI3980000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.