Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ MK 1903
GREENER_CHOICESDP

Catalog No. 462210 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-221-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 46-221-0 Supplier R&D Systems™ Supplier No. 4622/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective GPR109A (HCA2) agonist

Potent and selective hydroxycarboxylic acid receptor 2 (HCA2, GPR109A) full agonist; exhibits greater potency than niacin in a whole cell HTRF-cAMP assay (EC50 values are 12.9 and 51 nM respectively). Exhibits no binding at the GRP109B receptor.

Chemical Identifiers

CAS 1268882-43-4
Molecular Formula C8H8N2O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 164.16
Physical Form Solid
Color White
Purity >98%
Molecular Weight (g/mol) 164.164
Chemical Name or Material MK 1903
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.