Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MK 212 hydrochloride, Tocris Bioscience™

Catalog No. p-4785118
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-HT2C agonist

5-HT receptor agonist; displays selectivity for 5-HT2C over 5-HT2A (IC50 values are 0.028 and 0.42 μM for human 5-HT2C and 5-HT2A receptors expressed in HEK293 cells respectively).

Chemical Identifiers

CAS 64022-27-1
Molecular Formula C8H12Cl2N4
Molecular Weight (g/mol) 235.11
MDL Number MFCD04975444
InChI Key PFIZGLUIYAZQFU-UHFFFAOYSA-N
Synonym 2-chloro-6-1-piperazinyl pyrazine, 6-chloro-2-1-piperazinyl pyrazine, 2-chloro-6-piperazin-1-yl pyrazine, pyrazine, 2-chloro-6-1-piperazinyl, tocris-0941, biomol-nt_000130, d02esw, gtpl165, 1-6-chloro-2-pyrazinyl piperazine, 6-chloro-2-1-piperazinyl-pyrazine
PubChem CID 107992
IUPAC Name hydrogen 2-chloro-6-(piperazin-1-yl)pyrazine chloride
SMILES [H+].[Cl-].ClC1=CN=CC(=N1)N1CCNCC1

Specifications

Quantity 10 mg
Formula Weight 235.12
Percent Purity >99%
Chemical Name or Material MK 212 hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.