Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MK 212 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
5-HT receptor agonist; displays selectivity for 5-HT2C over 5-HT2A (IC50 values are 0.028 and 0.42 μM for human 5-HT2C and 5-HT2A receptors expressed in HEK293 cells respectively).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 235.12 |
Percent Purity | >99% |
Chemical Name or Material | MK 212 hydrochloride |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction