Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MK 2206 dihydrochloride
GREENER_CHOICESDP

Catalog No. 785010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-501-0 10 mg
78-505-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 78-501-0 Supplier Tocris Bioscience™ Supplier No. 7850/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective allosteric Akt1, Akt2 and Akt3 inhibitor

MK 2206 dihydrochloride is a potent and selective allosteric Akt inhibitor (IC50 values are 5 nM, 12 nM, and 65 nM for Akt1, Akt2, and Akt3, respectively). MK 2206 requires the Pleckstrin homology domain for its activity, and exhibits no inhibitory activity in a panel of 250 tested protein kinases. MK 2206 induces growth inhibition of different cancer cell lines (IC50 in the range 3.4 and 28.6 μmol/L) and enhances antitumor efficacy of a range of standard chemotherapeutics. It synergistically inhibits cell proliferation of human lung and breast cancer cells in combination with Erlotinib (Cat. No. 7194) by suppressing both the Ras/Erk and PI 3-K pathways in vitro and in vivo. MK 2206 can be used to block the IL-22 - Akt signaling pathway for studying paneth cell differentiation.

Specifications

Chemical Name or Material 8-[4-(1-Aminocyclobutyl)phenyl]-9-phenyl-1,2,4-triazolo[3,4-f][1,6]naphthyridin-3(2H)-one dihydrochloride
CAS 1032350-13-2
Quantity 10 mg
Target Akt (Protein Kinase B) Inhibitors
Molecular Formula C25H21N5O.2HCl
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.