Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ (+)-MK 801 maleate
GREENER_CHOICESDP

Catalog No. 092450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-245-0 50 mg
09-241-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 09-245-0 Supplier R&D Systems™ Supplier No. 0924/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-competitive NMDA antagonist; acts at ion channel site

(+)-MK 801 maleate is a high affinity (Ki = 37.2 nM), selective and non-competitive NMDA receptor antagonist. (+)-MK 801 maleate acts by binding to a site located within the NMDA associated ion channel and thus prevents Ca2+ flux. It is an effective anti-ischemic agent in several animal models. (+)-MK 801 maleate increases motor activity in rat models. (+)-MK 801 maleate inhibits proliferation and increases apoptosis in hippocampal neural stem cells (NSCs). (+)-MK 801 maleate induces schizophrenia-like symptoms in rodents.

(-)-enantiomer also available.

Chemical Identifiers

CAS 77086-22-7
Molecular Formula C20H19NO4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 337.37
Purity >99%
Molecular Weight (g/mol) 337.375
Chemical Name or Material (+)-MK 801 maleate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.