Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mKate Mouse anti-Tag, Clone: OTI1G5, TrueMAB™, Origene
SDP

Catalog No. 502241678
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-224-1678 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-224-1678 Supplier OriGene Supplier No. TA180091S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

mKate Monoclonal antibody specifically detects mKate in Tag samples. It is validated for Western Blot

Specifications

Antigen mKate
Applications Western Blot
Classification Monoclonal
Clone OTI1G5
Concentration 0.5-1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Alias mKate
Host Species Mouse
Immunogen Recombinant protein and recognizes mKate.
Purification Method Protein G
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.