Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mKate Rabbit anti-Tag, Polyclonal, liquid, OriGene
SDP

Catalog No. 501671080
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1080 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1080 Supplier OriGene Supplier No. TA150072
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

mKate fluorescent protein.

Specifications

Antigen mKate
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene Alias mKate
Host Species Rabbit
Immunogen Purified mKate protein expressed in E.coli
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.