Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKI67IP Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501566757
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6757 Supplier Bethyl Laboratories, Inc Supplier No. A304047A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

NIFK is a nucleolar protein that interacts with Ki67, a protein strongly associated with, and a specific marker of, cell proliferation. The marsupial counterpart of human NIFK was recently identified and suggests that NIFK may be involved in the organization of higher order chromatin structure. NIFK was found to interact with the forkhead associated domain of Ki67 in mitotic cells and the interaction is thought to be phosphorylation-dependent.

Specifications

Antigen MKI67IP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene NIFK
Gene Accession No. Q9BYG3
Gene Alias hNIFK, MKI67 (FHA domain) interacting nucleolar phosphoprotein, MKI67IP, Nopp34, nucleolar phosphoprotein Nopp34, nucleolar protein interacting with the FHA domain of pKi-67
Gene Symbols NIFK
Host Species Rabbit
Immunogen Between 243 and 293
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84365
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.