Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKK4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501563143
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3143 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3143 Supplier Bethyl Laboratories, Inc Supplier No. A302657AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

MAP2K4 (SEK1) is a dual specificity protein kinase that belongs to the Ser/Thr protein kinase family. This kinase is a direct activator of MAP kinases in response to various environmental stresses or mitogenic stimuli. It has been shown to activate MAPK8/JNK1, MAPK9/JNK2, and MAPK14/p38, but not MAPK1/ERK2 or MAPK3/ERK3. MAP2K4 is phosphorylated and activated by MAP3K1/MEK.

Specifications

Antigen MKK4
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene MAP2K4
Gene Accession No. P45985
Gene Alias c-Jun N-terminal kinase kinase 1, JNK-activated kinase 1, JNK-activating kinase 1, JNKK, JNKK1, MAP kinase kinase 4, MAPK/ERK kinase 4, MAPKK 4, MAPKK4, MEK 4, MEK4, MKK4, PRKMK4, SAPK kinase 1, SAPK/ERK kinase 1, SAPKK1, SAPKK-1, SEK1, SERK1, SKK1, stress-activated protein kinase kinase 1
Gene Symbols MAP2K4
Host Species Rabbit
Immunogen Between 50 and 100
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6416
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.