Learn More
MKK4 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302658A

Description
The recommended shelf life for this product is 1 year from date of receipt.
MAP2K4 (SEK1) is a dual specificity protein kinase that belongs to the Ser/Thr protein kinase family. This kinase is a direct activator of MAP kinases in response to various environmental stresses or mitogenic stimuli. It has been shown to activate MAPK8/JNK1, MAPK9/JNK2, and MAPK14/p38, but not MAPK1/ERK2 or MAPK3/ERK3. MAP2K4 is phosphorylated and activated by MAP3K1/MEK.Specifications
MKK4 | |
Polyclonal | |
Unconjugated | |
MAP2K4 | |
c-Jun N-terminal kinase kinase 1, JNK-activated kinase 1, JNK-activating kinase 1, JNKK, JNKK1, MAP kinase kinase 4, MAPK/ERK kinase 4, MAPKK 4, MAPKK4, MEK 4, MEK4, MKK4, PRKMK4, SAPK kinase 1, SAPK/ERK kinase 1, SAPKK1, SAPKK-1, SEK1, SERK1, SKK1, stress-activated protein kinase kinase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26398, 6416 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P45985, P47809 | |
MAP2K4 | |
Between 349 and 399 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.