Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MKK6/MEK6, p Ser207 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$268.00 - $426.00
Specifications
Antigen | MKK6/MEK6 (p Ser207) |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017861
![]() |
Novus Biologicals
NB100820500.025ML |
0.025 mL |
Each for $268.00
|
|
|||||
NB10082050
![]() |
Novus Biologicals
NB10082050 |
0.05 mL |
Each for $426.00
|
|
|||||
Description
MKK6/MEK6 Polyclonal specifically detects MKK6/MEK6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MKK6/MEK6 (p Ser207) | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 | |
Polyclonal | |
Rabbit | |
Angiogenesis, Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Phospho Specific | |
P52564 | |
5608 | |
IgG | |
Affinity Purified |
1.0 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
RUO | |
Rat, Human, Mouse | |
MAP kinase kinase 6, MAPK/ERK kinase 6, MAPKK6dual specificity mitogen-activated protein kinase kinase 6, MEK6MAPKK 6, mitogen-activated protein kinase kinase 6, MKK6MEK 6, PRKMK6protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6), SAPKK3EC 2.7.12.2 | |
MAP2K6 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title