Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MKKS Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299025MFV450
Description
MKKS Polyclonal antibody specifically detects MKKS in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
MKKS | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
Bardet-Biedl syndrome 6 protein, BBS6MKS, HMCS, KMS, McKusick-Kaufman syndrome, McKusick-Kaufman/Bardet-Biedl syndromes putative chaperonin | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human MKKS. | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Immunology, Signal Transduction | |
8195 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction