Learn More
Invitrogen™ MKKS Polyclonal Antibody

Description
Immunogen sequence: ALESVVGSLE HDGGEILTDM KYGHLWSVQA DSPCVANWPD LLSQCGCGLY NSQEELNWSF LRSTRRPFVP QSCLPHEAVG SASNLTL Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 74%.
Specifications
Specifications
| Antigen | MKKS |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MKKS |
| Gene Accession No. | Q9NPJ1 |
| Gene Alias | 1300013E18Rik; AI463362; AI957237; Bardet-Biedl syndrome 6 protein; BBS6; HMCS; KMS; McKusick-Kaufman syndrome; McKusick-Kaufman syndrome protein; McKusick-Kaufman syndrome protein; MKKS protein; McKusick-Kaufman/Bardet-Biedl syndromes putative chaperonin; MKKS; MKS; protein Bbs6 homolog |
| Gene Symbols | MKKS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.