Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MKKS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560653
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560653 Supplier Invitrogen™ Supplier No. PA560653
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSTALKVALF CTTLSGDTSD TGEGTVVVSY GVSLENAVLD QLLNLGRQLI SDHVDLVLCQ KVIHPSLKQF LNMHRIIAID RIGVTLMEPL TK Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 80%.

Probable molecular chaperone. Assists the folding of proteins upon ATP hydrolysis. As part of the BBS/CCT complex may play a role in the assembly of BBSome, a complex involved in ciliogenesis regulating transports vesicles to the cilia. May play a role in protein processing in limb, cardiac and reproductive system development. May play a role in cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKKS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MKKS
Gene Accession No. Q9NPJ1
Gene Alias 1300013E18Rik; AI463362; AI957237; Bardet-Biedl syndrome 6 protein; BBS6; HMCS; KMS; McKusick-Kaufman syndrome; McKusick-Kaufman syndrome protein; McKusick-Kaufman syndrome protein; MKKS protein; McKusick-Kaufman/Bardet-Biedl syndromes putative chaperonin; MKKS; MKS; protein Bbs6 homolog
Gene Symbols MKKS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MKKS. Recombinant protein control fragment (Product #RP-99409).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8195
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.