Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mkl1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP180259
Description
Mkl1 Polyclonal specifically detects Mkl1 in Mouse samples. It is validated for Western Blot.Specifications
Mkl1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
AMKL, KIAA1438, MALSAP and coiled-coil domain, megakaryoblastic leukemia (translocation) 1, Megakaryoblastic leukemia 1 protein, Megakaryocytic acute leukemia protein, MKL/myocardin-like protein 1, Myocardin-related transcription factor A, RNA-binding motif protein 15/megakaryoblastic leukemia-1 fusion protein | |
Rabbit | |
Protein A purified | |
RUO | |
57591 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_694629 | |
MRTFA | |
The immunogen is a synthetic peptide corresponding to a region of Mouse. Peptide sequence QPLSQPGFPAPGPPAQMDLEHPPQPPFATPTSLLKKEPPGYEETVTQQPK. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Mouse: 100%; Rat: 100%; Bovine: 85%; Canine: 85%; Equine: 85%; Pig: 85%; Rabbit: 85%; Human: 78%; Chicken: 76%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit, Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction